Learn More
Gibson Bioscience India Ink Control Kit
Description
Requires
The usual microbiological equipment such as slides, coverslips, microscope, biohazard bags, etc. are needed for procedures involving the use of this product.
India Ink wet mounts are used for the visualization of encapsulated microorganisms, especially Cryptococcus neoformans.
- India Ink is added to each positive and negative suspension on a glass slide and observed under a microscope.
- The positive control provides an encapsulated suspension of Cryptococcus neoformans to be utilized when performing India Ink procedures
- The negative control provides a non-encapsulated suspension of Candida sp. to be utilized when performing India Ink
- Positive: The polysaccharide capsules will exclude the particles of ink, and the capsules will appear as clear halos around the organisms on semiopaque backgrounds
- Negative: No capsules present, no clear halos observed
Specifications
Specifications
| Type | India Ink Positive Control Kit |
| Quantity | 20 Tests |
| Format | Dropper |
| For Use With (Application) | Visualization of encapsulated microorganisms |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.