Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GID4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569986
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569986 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569986 Supplier Invitrogen™ Supplier No. PA569986
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 93%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.

The multiprotein Mediator complex is a coactivator required for activation of RNA polymerase II transcription by DNA bound transcription factors. The protein encoded by this gene is thought to be a subunit of the Mediator complex. This gene is located within the Smith-Magenis syndrome region on chromosome 17.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GID4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GID4
Gene Accession No. Q8IVV7
Gene Alias 4933439F18Rik; C17orf39; C19H17orf39; C5H17orf39; GID complex subunit 4; GID complex subunit 4 homolog; GID complex subunit 4 homolog; glucose-induced degradation protein 4 homolog; GID complex subunit 4, VID24 homolog; GID complex subunit 4, VID24 homolog (S. cerevisiae); GID4; Glucose-induced degradation protein 4 homolog; im:7147459; LOC509503 protein; LOW QUALITY PROTEIN: glucose-induced degradation protein 4 homolog; vacuolar import and degradation 24; vacuolar import and degradation protein 24 homolog; VID24
Gene Symbols GID4
Host Species Rabbit
Immunogen synthetic peptide directed towards the C terminal region of human GID4.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79018
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.