Learn More
Epredia™ Giemsa Stain
Catalog No. p-4530209 Shop All Epredia Products
Click to view available options
Quantity:
32 oz.
1 gal.
This item is not returnable. View return policy
This item is not returnable. View return policy
Take advantage of an excellent classical blood film stain used on peripheral blood smears and bone marrow with the Epredia™ Giemsa Stain.
  • Erythrocytes stain pink with platelets exhibiting a very light, pale hue
  • Leukocyte nuclear chromatin stains magenta-purple
  • Monocyte cytoplasm stains the classical “ground glass” blue
  • Lymphocyte cytoplasm stains clear, sky blue
Specifications
Quantity 32 oz.
Type Giemsa Stain
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.