Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Giemsa Stain, Stock, for Staining of Blood, Tissue, and Malarial Parasites in Blood, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
120 mL
500 mL
Description
- Components: Giemsa Stain, Glycerin, Methanol
- Shelf life: 24 months
Used for blood staining and Malarial parasite staining applications
Specifications
Specifications
Quantity | 500 mL |
Type | Giemsa Stain |
Packaging | Natural Poly Bottle |
CAS | 56-81-5, 67-56-1, 51811-82-6 |
Product Type | Giemsa Stain |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction