Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GIF Antibody (012), mFluor Violet 500 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290226MFV500
Description
GIF Monoclonal antibody specifically detects GIF in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
GIF | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
2694 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunoprecipitation | |
012 | |
Western Blot, Immunoprecipitation | |
gastric intrinsic factor (vitamin B synthesis), IFIntrinsic factor, IFMHgastric intrinsic factor, INF, TCN3 | |
Rabbit immunized with purified, recombinant Human GIF (Uniprot#: P27352-1, Met1-Tyr417) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction