Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GIF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597783
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597783 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597783 Supplier Invitrogen™ Supplier No. PA597783
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GIF Polyclonal Antibody for Western Blot, IHC (P), ELISA

Promotes absorption of the essential vitamin cobalamin (Cbl) in the ileum. After interaction with CUBN, the GIF-cobalamin complex is internalized via receptor-mediated endocytosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GIF
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene CBLIF
Gene Accession No. P27352, P52787
Gene Alias AV073125; Cblif; Cobalamin binding intrinsic factor; gastric intrinsic factor; gastric intrinsic factor (vitamin B synthesis); gastric intrinsic factor-like; Gif; IF; IFMH; INF; intestinal cobalamin binding protein; intrinsic factor; Pancreatic intrinsic factor; TCN3
Gene Symbols CBLIF
Host Species Rabbit
Immunogen Recombinant Human Cobalamin binding intrinsic factor protein (19-417AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14603, 2694
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.