Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GIF Recombinant Rabbit Monoclonal Antibody (016)
GREENER_CHOICE

Catalog No. PIMA530462
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530462 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530462 Supplier Invitrogen™ Supplier No. MA530462
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human Gastric intrinsic factor/GIF.

Promotes absorption of the essential vitamin cobalamin (Cbl) in the ileum. After interaction with CUBN, the GIF-cobalamin complex is internalized via receptor-mediated endocytosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GIF
Applications ELISA
Classification Recombinant Monoclonal
Clone 016
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene CBLIF
Gene Accession No. P27352
Gene Alias AV073125; Cblif; Cobalamin binding intrinsic factor; gastric intrinsic factor; gastric intrinsic factor (vitamin B synthesis); gastric intrinsic factor-like; Gif; IF; IFMH; INF; intestinal cobalamin binding protein; intrinsic factor; Pancreatic intrinsic factor; TCN3
Gene Symbols CBLIF
Host Species Rabbit
Immunogen Recombinant Human Gastric intrinsic factor/GIF Protein (Met1-Tyr417).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2694
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.