Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gigaxonin Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP149923AF532
Description
Gigaxonin Polyclonal antibody specifically detects Gigaxonin in Human samples. It is validated for Western BlotSpecifications
Gigaxonin | |
Polyclonal | |
Western Blot | |
FLJ38059, GAN1KLHL16Kelch-like protein 16, giant axonal neuropathy (gigaxonin), gigaxonin | |
The immunogen for this product maps to a region between residue 50 and 100 of human Gigaxonin using the numbering given in entry NP_071324.1 (GeneID 8139). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
8139 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction