Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gigaxonin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP149923
Description
Gigaxonin Polyclonal specifically detects Gigaxonin in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative).Specifications
Gigaxonin | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
GAN | |
The immunogen for this product maps to a region between residue 50 and 100 of human Gigaxonin using the numbering given in entry NP_071324.1 (GeneID 8139). | |
100 μL | |
Primary | |
Human | |
IgG |
Immunoprecipitation, Western Blot | |
1.0 mg/mL | |
Immunoprecipitation 2-5 ug/mg lysate, Western Blot (Negative) | |
FLJ38059, GAN1KLHL16Kelch-like protein 16, giant axonal neuropathy (gigaxonin), gigaxonin | |
Rabbit | |
Affinity Purified | |
RUO | |
8139 | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction