Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gigaxonin Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981040
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1040 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1040 Supplier Bioss Supplier No. BS11025R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a member of the cytoskeletal BTB/kelch repeat family. The encoded protein plays a role in neurofilament architecture and is involved in mediating the ubiquitination and degradation of some proteins. Defects in this gene are a cause of giant axonal neuropathy.

Specifications

Antigen Gigaxonin
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GAN
Gene Accession No. Q9H2C0
Gene Alias A330045G18; BOS_16975; GAN; GAN1; giant axonal neuropathy; giant axonal neuropathy (gigaxonin); Gigaxonin; kelch-like family member 16; Kelch-like protein 16; KLHL16
Gene Symbols GAN
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Gigaxonin.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8139
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.