Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GIGYF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554302
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554302 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554302 Supplier Invitrogen™ Supplier No. PA554302
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DTLEAKEFAK QFLERRAKQK ASQQRQQQQE AWLSSASLQT AFQANHSTKL GPGEGSKAKR RALMLHSDPS ILGYSL Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 86%.

May act cooperatively with GRB10 to regulate tyrosine kinase receptor signaling. May increase IGF1 receptor phosphorylation under IGF1 stimulation as well as phosphorylation of IRS1 and SHC1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GIGYF1
Applications Immunocytochemistry, Immunohistochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GIGYF1
Gene Accession No. O75420
Gene Alias CDS2; GIGYF1; GRB10 interacting GYF protein 1; GRB10-interacting GYF protein 1; GYF domain containing 1; GYF1; PERQ amino acid rich, with GYF domain 1; PERQ amino acid-rich with GYF domain-containing protein 1; PERQ1; postmeiotic segregation increased 2-like 12; PP3360
Gene Symbols GIGYF1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GIGYF1. Recombinant protein control fragment (Product #RP-92754).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64599
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.