Learn More
GIGYF1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304132A

Description
The recommended shelf life for this product is 1 year from date of receipt.
May act cooperatively with GRB10 to regulate tyrosine kinase receptor signaling. May increase IGF1 receptor phosphorylation under IGF1 stimulation as well as phosphorylation of IRS1 and SHC1.Specifications
GIGYF1 | |
Polyclonal | |
Unconjugated | |
GIGYF1 | |
PERQ amino acid-rich with GYF domain-containing protein 1, GRB10-interacting GYF protein 1, GIGYF1, CDS2, PERQ1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64599 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
O75420 | |
GIGYF1 | |
Between 350 and 400 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.