Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GIGYF2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563793
Description
Immunogen sequence: QQYAQVLAQQ QKAALSSQQQ QQLALLLQQF QTLKMRISDQ NIIPSVTRSV SVPDTGSIWE LQPTASQPTV WEGGSVWDLP L Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
The function of this protein remains unknown.
Specifications
GIGYF2 | |
Polyclonal | |
Unconjugated | |
GIGYF2 | |
2610016F01Rik; A830080H02Rik; AI852361; AW259676; BC006835; GIGYF2; GRB10 interacting GYF protein 2; GRB10 interacting GYF protein 2; GRB10-interacting GYF protein 2; GRB10-interacting GYF protein 2; GYF2; KIAA0642; LOW QUALITY PROTEIN: PERQ amino acid-rich with GYF domain-containing protein 2; mKIAA0642; PARK11; Parkinson disease (autosomal recessive, early onset) 11; PERQ amino acid rich, with GYF domain 3; PERQ amino acid-rich with GYF domain-containing 2-like protein; PERQ amino acid-rich with GYF domain-containing protein 2; PERQ amino acid-rich with GYF domain-containing protein 2; GRB10-interacting GYF protein 2; PERQ2; PERQ3; rCG23949-like; TNRC15; trinucleotide repeat containing 15; trinucleotide repeat-containing gene 15 protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26058 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6Y7W6 | |
GIGYF2 | |
Recombinant protein corresponding to Human GIGYF2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction