Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GIGYF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565646
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565646 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565646 Supplier Invitrogen™ Supplier No. PA565646
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QQYAQVLAQQQ KAALSSQQQQ QLALLLQQFQ TLKMRISDQN IIPSVTRSVS VPDTGSIWEL QPTASQPTVW EGGSVWDLPL Highest antigen sequence identity to the following orthologs - mouse 95%, rat 95%.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GIGYF2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GIGYF2
Gene Accession No. Q6Y7W6
Gene Alias 2610016F01Rik; A830080H02Rik; AI852361; AW259676; BC006835; GIGYF2; GRB10 interacting GYF protein 2; GRB10 interacting GYF protein 2; GRB10-interacting GYF protein 2; GRB10-interacting GYF protein 2; GYF2; KIAA0642; LOW QUALITY PROTEIN: PERQ amino acid-rich with GYF domain-containing protein 2; mKIAA0642; PARK11; Parkinson disease (autosomal recessive, early onset) 11; PERQ amino acid rich, with GYF domain 3; PERQ amino acid-rich with GYF domain-containing 2-like protein; PERQ amino acid-rich with GYF domain-containing protein 2; PERQ amino acid-rich with GYF domain-containing protein 2; GRB10-interacting GYF protein 2; PERQ2; PERQ3; rCG23949-like; TNRC15; trinucleotide repeat containing 15; trinucleotide repeat-containing gene 15 protein
Gene Symbols GIGYF2
Host Species Rabbit
Immunogen Recombinant Human GIGYF2. Recombinant protein control fragment (Product #RP-99707).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26058
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.