Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GIGYF2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA588331
Description
Immunogen sequence: VDEGEECSDS EGSHNEEAKE PDKTNKKEGE KTDRVGVASE ETPQTSSSSA RPGTPSDHQS QEASQFERKD EPKTEQTEKA EEETRMENSL PAKVPSRGDE MVADVQQPLS QIPSDTASPL LILPPPVPNP SPTLRPVETP VVGAPGMGSV S; Positive Samples: 293T, HeLa, Jurkat, Raji, LO2.
The function of this protein remains unknown.
Specifications
GIGYF2 | |
Polyclonal | |
Unconjugated | |
GIGYF2 | |
2610016F01Rik; A830080H02Rik; AI852361; AW259676; BC006835; GIGYF2; GRB10 interacting GYF protein 2; GRB10 interacting GYF protein 2; GRB10-interacting GYF protein 2; GRB10-interacting GYF protein 2; GYF2; KIAA0642; LOW QUALITY PROTEIN: PERQ amino acid-rich with GYF domain-containing protein 2; mKIAA0642; PARK11; Parkinson disease (autosomal recessive, early onset) 11; PERQ amino acid rich, with GYF domain 3; PERQ amino acid-rich with GYF domain-containing 2-like protein; PERQ amino acid-rich with GYF domain-containing protein 2; PERQ amino acid-rich with GYF domain-containing protein 2; GRB10-interacting GYF protein 2; PERQ2; PERQ3; rCG23949-like; TNRC15; trinucleotide repeat containing 15; trinucleotide repeat-containing gene 15 protein | |
Rabbit | |
Affinity Chromatography | |
RUO | |
100362458, 227331, 26058 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.69 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q6Y7W6, Q6Y7W8 | |
GIGYF2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 350-500 of human GIGYF2 (NP_0010966171). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction