Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GIGYF2 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501565909
Encompass_Preferred
Change view
Click to view available options
20 μl; Unlabeled
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
50-156-5909 20 μl; Unlabeled
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-5909 Supplier Bethyl Laboratories, Inc Supplier No. A303732AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse

The function of this protein remains unknown.

Specifications

Antigen GIGYF2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene GIGYF2
Gene Accession No. Q6Y7W6
Gene Alias GRB10-interacting GYF protein 2, GYF domain containing 2, GYF2, KIAA0642, PARK11, Parkinson disease (autosomal recessive, early onset) 11, PERQ amino acid rich, with GYF domain 2, PERQ amino acid rich, with GYF domain 3, PERQ2, PERQ3, TNRC15, trinucleotide repeat containing 15, trinucleotide repeat-containing gene 15 protein
Gene Symbols GIGYF2
Host Species Rabbit
Immunogen Between 1249 and 1299
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26058
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.