Learn More
Epredia™ Gill™ Hematoxylin 1, 2, and 3
Catalog No. p-4530210
Click to view available options
Quantity:
1 pt.
1 gal.
Type:
Gill 1 Hematoxylin
Gill 2 Hematoxylin
Gill 3 Hematoxylin
With three strengths available, the Epredia™ Gill™ 1, 2, 3 Hematoxylin allows complete control of staining intensities.
Gill Hematoxylins are progressive nuclear stains.
  • Gill 1 is recommended for use in cytology
  • Gill 2 has applications in histology and cytology
  • Gill 3 is used in histology
  • All are pre-mixed and ready-to-use
Specifications
Type Gill 1 Hematoxylin
Quantity 1 pt.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.