Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gilson™ PIPETMANTM EcoPackTM Tips
GREENER_CHOICE

Catalog No. p-4330374 Shop All Gilson Products
Click to view available options
Volume (Metric):
0.1 to 10 μL
0.1 to 20 μL
100 to 1000 μL
100 to 1200 μL
2 to 200 μL
20 to 300 μL

Ensures optimal performance of PIPETMAN pipettes across the full volume range. GilsonTM PIPETMANTM EcoPackTM Tips are fully autoclavable.

  • Optimized geometry and pure polypropylene material ensure low retention and naturally hydrophobic surface.
  • Manufactured in a cleanroom and hence no risk of contamination.
  • Full traceability from the mold to the bench guaranteed.
  • Volume graduations make it easy to visually check the volume aspirated.
  • Requires low tip-mounting force.
  • Easy identification with color-coded packaging.
  • Bulk Packs are sealed with a tamper-proof strip which can easily be opened for quick access.
  • Bulk Packs are closed using the rapid-sealing strip to prevent contamination and loss of tips.
  • Each EcoPack box contains 10,000 tips (10 bags of 1000 tips).

Specifications

Volume (Metric) 100 to 1200 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Model D1200
Length (English) 15.63 in.
Length (Metric) 39.7 cm
Diameter (Metric) Outer .03 in
Compatibility PIPETMAN Pipettes
Format Bagged
Tip Style Universal
Quantity 10000
Product Type Pipet Tips
Color Code Blue
Disposable Single-use
For Use With (Equipment) Gilson Pipetman
Graduated Yes
Product Line Pipetman EcoPack
Material Polypropylene
Graduations 100 μL, 1000 μL
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.