Learn More
Gilson™ PIPETMAN™ Neo Pipets
Catalog No. F144566G Shop All Gilson Products
Click to view available options
Volume (Metric):
100 to 1000 μL
20 to 200 μL
This item is not returnable. View return policy
This item is not returnable. View return policy
Fits most brands of tips. Gilson PIPETMAN™ Neo Pipets are the next generation of the PIPETMAN Classic™ with lower spring and ejection forces.
  • Redesigned tip ejector and ejection springs reduces tip ejection forces
  • Unique clip ejector fits securely into the body of the pipette to avoid slipping over time
  • PVDF handle limits the transfer of heat from hand to the pipette and is resistant to aggressive chemicals
  • Stainless steel tip ejector resists corrosion and offers a smooth tip ejection stroke
  • Color-coded push-button for ease of identification
  • Unique tip holder allows for a secure fit of most brands of tip
  • Models P2N and P10N incorporate dual position tip ejectors, allowing the use of D10 (short) and DL10 (long)
Order Info
Trade in program available, please contact your Fisher Scientific Sales representative for more details.
Specifications
Accuracy ±3, ±4, ±8μL
Certifications/Compliance ISO8655
Compatible Tips D1000, DF1000ST, DF1200ST
Model P1000N
Number of Channels 1
Product Line Pipetman Neo
Type Pipette
Volume (Metric) 100 to 1000 μL
Finger Style Ergonomic
Includes Color coded push button, PVDF handle, Stainless Steel tip ejectors, universal tip holder
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.