Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gilson™ Safe Aspiration Station
Click to view available options
Product Type:
Tubing Connector
Description
- Closed, stand-alone unit designed to prevent contamination hazards
- Compact design with adjustable speed allowing quick aspiration
- Always ready to use by simply pressing the handle button
- Handle accommodates different types of adaptors to aspirate from a variety of vessels
- Convenient coupling for quick bottle emptying
- In-line hydrophobic filter traps aerosols
- Connect two handles to the station with the Y-shape connector
- Motor starts only when the vacuum pressure drops
- Level detection prevents overflow
Specifications
Specifications
Product Type | Tubing Connector |
Color | Clear |
For Use With (Equipment) | Gilson safe aspiration station |
Quantity | 1 Ea. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction