Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GILT/IFI30 Antibody (003), Alexa Fluor™ 647, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290133AF647
Description
GILT/IFI30 Monoclonal specifically detects GILT/IFI30 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
GILT/IFI30 | |
Monoclonal | |
Alexa Fluor 647 | |
EC 1.8, Gamma-interferon-inducible protein IP-30, GILTgamma-interferon-inducible lysosomal thiol reductase, IFI-30, interferon, gamma-inducible protein 30, IP30interferon gamma-inducible protein 30 preproprotein, Legumaturain, MGC32056 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human GILT/IFI30 (P13284; Met1-Lys243). | |
0.1 mL | |
Stem Cell Markers | |
10437 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
3 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction