Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GILT/IFI30 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300027AF405
Description
GILT/IFI30 Polyclonal specifically detects GILT/IFI30 in Human samples. It is validated for Western Blot, ELISA.Specifications
GILT/IFI30 | |
Polyclonal | |
Western Blot, ELISA | |
EC 1.8, Gamma-interferon-inducible protein IP-30, GILTgamma-interferon-inducible lysosomal thiol reductase, IFI-30, interferon, gamma-inducible protein 30, IP30interferon gamma-inducible protein 30 preproprotein, Legumaturain, MGC32056 | |
Produced in rabbits immunized with purified, recombinant GILT/IFI30 (P13284; Met1-Lys243). | |
0.1 mL | |
Stem Cell Markers | |
10437 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction