Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GILT/IFI30 Rabbit anti-Human, Clone: 30, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP29013450UL
Description
GILT/IFI30 Monoclonal specifically detects GILT/IFI30 in Human samples. It is validated for ELISA.Specifications
GILT/IFI30 | |
Monoclonal | |
Unconjugated | |
0.2 um filtered solution in PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
30 | |
ELISA 1:5000-1:10000 | |
EC 1.8, Gamma-interferon-inducible protein IP-30, GILTgamma-interferon-inducible lysosomal thiol reductase, IFI-30, interferon, gamma-inducible protein 30, IP30interferon gamma-inducible protein 30 preproprotein, Legumaturain, MGC32056 | |
Recombinant Human GILT/IFI30 protein | |
50 μg | |
Stem Cell Markers | |
10437 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction