Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GilZ Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP241210
Description
GilZ Polyclonal specifically detects GilZ in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
GilZ | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
TSC22D3 | |
Antibody was raised against a 16 amino acid peptide near the amino terminus of human TSC22D3. The immunogen is located within amino acids 40 - 90 of TSC22D3. Amino Acid Squence: ENLNTDKLNSIMRQDS | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Western Blot 1 - 2 μg/mL, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:10-1:500 | |
Delta sleep-inducing peptide immunoreactor, DSIP-immunoreactive leucine zipper protein, DSIP-immunoreactive peptide, GILZDKFZp313A1123, Glucocorticoid-induced leucine zipper protein, Protein DIP, TSC22 domain family protein 3, TSC22 domain family, member 3, TSC-22 related protein, TSC-22-like protein, TSC-22-related protein, TSC-22Rimmunoreactor | |
Rabbit | |
23 kDa | |
0.1 mg | |
Cancer | |
1831 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction