Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GilZ Antibody (CFMKG15), DyLight 350, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP200143UV
Description
GilZ Monoclonal specifically detects GilZ in Mouse samples. It is validated for Western Blot, Flow Cytometry.Specifications
| GilZ | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| TSC22D3 | |
| The immunogen for this antibody is GILZ. | |
| 100 μL | |
| Cancer | |
| 1831 | |
| Store at 4C in the dark. | |
| IgG2a κ |
| Western Blot, Flow Cytometry | |
| CFMKG15 | |
| Western Blot, Flow Cytometry | |
| Delta sleep-inducing peptide immunoreactor, DSIP-immunoreactive leucine zipper protein, DSIP-immunoreactive peptide, GILZDKFZp313A1123, Glucocorticoid-induced leucine zipper protein, Protein DIP, TSC22 domain family protein 3, TSC22 domain family, member 3, TSC-22 related protein, TSC-22-like protein, TSC-22-related protein, TSC-22Rimmunoreactor | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction