Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GilZ Antibody (CFMKG15), FITC, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP200143F
Description
GilZ Monoclonal specifically detects GilZ in Mouse samples. It is validated for Flow Cytometry.Specifications
GilZ | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
TSC22D3 | |
The immunogen for this antibody is GILZ. | |
100 μL | |
Cancer | |
1831 | |
Store at 4C in the dark. | |
IgG2a κ |
Flow Cytometry | |
CFMKG15 | |
Flow Cytometry | |
Delta sleep-inducing peptide immunoreactor, DSIP-immunoreactive leucine zipper protein, DSIP-immunoreactive peptide, GILZDKFZp313A1123, Glucocorticoid-induced leucine zipper protein, Protein DIP, TSC22 domain family protein 3, TSC22 domain family, member 3, TSC-22 related protein, TSC-22-like protein, TSC-22-related protein, TSC-22Rimmunoreactor | |
Rat | |
Protein A or G purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction