Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GILZ Monoclonal Antibody (CFMKG15), PE, eBioscience™
GREENER_CHOICE

Catalog No. 5010770
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-107-70 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-107-70 Supplier Invitrogen™ Supplier No. 12403382
Add to Cart
Add to Cart

Rat Monoclonal Antibody

GILZ is a leucine-zipper-containing protein that is induced by glucocorticoids. It is expressed in mast cells, monocytes, macrophages, dendritic cells, and T cells. GILZ has been shown to inhibit production of IL-2 as well as TCR-driven upregulation of FasL. GILZ mediates these changes in transcription, at least in part, by inhibiting DNA-binding of the transcription factors AP-1 and NF-kappaB. GILZ has also been shown to mediate anti-inflammatory signals in macrophages and dendritic cells by interfering with NF-kappaB-mediated signaling. Although GILZ has been reported to localize to the nucleus, more recent reports as well as testing at eBioscience have observed predominantly cytoplasmic staining. Applications Reported: This CFMKG15 antibody has been reported for use in intracellular staining followed by flow cytometric analysis and immunocytochemistry. Applications Tested: This CFMKG15 antibody has been tested by intracellular flow cytometric analysis of dexamethasone-treated mouse lymph node cells and human peripheral blood cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. For intracellular staining with CFMKG15, it is recommended t...

The TSC22 domain family member 3 protein (TSC22D3) is a leucine zipper protein that functions as a transcriptional regulator. The expression of TSC22D3 is stimulated by glucocorticoids and IL-10 and is thought to play a key role in the anti-inflammatory and immunosuppressive effects of these molecules. TSC22D3 can physically interact with and inhibit the activities of key inflammatory signaling mediators such NF-Kappa-B and AP-1. TSC22D3 functions as a transcriptional co-activator for various nuclear receptors and NF-Kappa-B. It has also been shown to be involved in the differentiation of mesenchymal stem cells towards osteoblasts and bone formation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GILZ
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone CFMKG15
Concentration 0.2 mg/mL
Conjugate PE
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene TSC22D3
Gene Accession No. Q99576, Q9Z2S7
Gene Alias delta sleep inducing peptide, immunoreactor; delta sleep-inducing peptide immunoreactor; DIP; Dsip1; DSIPI; DSIP-immunoreactive leucine zipper protein; DSIP-immunoreactive peptide; GILZ; glucocorticoid-induced leucine zipper protein; long glucocorticoid-induced leucine zipper protein; Protein DIP; Tilz3; TSC22 domain family 3; TSC22 domain family member 3; TSC22 domain family protein 3; TSC22 domain family, member 3; TSC-22 related protein; TSC22D3; TSC-22-like protein; TSC-22R; TSC22-related inducible leucine zipper 3; TSC-22-related protein; TSC22-related-inducible leucine zipper 3
Gene Symbols TSC22D3
Host Species Rat
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14605, 1831
Target Species Human, Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.