Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GILZ Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534506
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534506 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534506 Supplier Invitrogen™ Supplier No. PA534506
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is human small intestine tissue lysate. PA5-34506 can be used with blocking peptide PEP-1549.

The TSC22 domain family member 3 protein (TSC22D3) is a leucine zipper protein that functions as a transcriptional regulator. The expression of TSC22D3 is stimulated by glucocorticoids and IL-10 and is thought to play a key role in the anti-inflammatory and immunosuppressive effects of these molecules. TSC22D3 can physically interact with and inhibit the activities of key inflammatory signaling mediators such NF-Kappa-B and AP-1. TSC22D3 functions as a transcriptional co-activator for various nuclear receptors and NF-Kappa-B. It has also been shown to be involved in the differentiation of mesenchymal stem cells towards osteoblasts and bone formation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GILZ
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene TSC22D3
Gene Accession No. Q99576, Q9Z2S7
Gene Alias delta sleep inducing peptide, immunoreactor; delta sleep-inducing peptide immunoreactor; DIP; Dsip1; DSIPI; DSIP-immunoreactive leucine zipper protein; DSIP-immunoreactive peptide; GILZ; glucocorticoid-induced leucine zipper protein; long glucocorticoid-induced leucine zipper protein; Protein DIP; Tilz3; TSC22 domain family 3; TSC22 domain family member 3; TSC22 domain family protein 3; TSC22 domain family, member 3; TSC-22 related protein; TSC22D3; TSC-22-like protein; TSC-22R; TSC22-related inducible leucine zipper 3; TSC-22-related protein; TSC22-related-inducible leucine zipper 3
Gene Symbols TSC22D3
Host Species Rabbit
Immunogen A 16 amino acid peptide near the amino terminus of human TSC22D3.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14605, 1831
Target Species Human, Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.