Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GIMAP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5101617
Description
Antibody detects endogenous levels of total GIMAP2.
This gene encodes a protein belonging to the GTP-binding superfamily and to the immuno-associated nucleotide (IAN) subfamily of nucleotide-binding proteins. In humans, the IAN subfamily genes are located in a cluster at 7q36.1.
Specifications
GIMAP2 | |
Polyclonal | |
Unconjugated | |
GIMAP2 | |
D15S9; GIMAP2; GTPase IMAP family member 2; GTPase, IMAP family member 2; HIMAP2; IAN12; IMAP2; immune-associated nucleotide-binding protein 12; immunity associated protein 2; immunity-associated protein 2; RNF63; ZNF127 | |
Rabbit | |
Affinity chromatography | |
RUO | |
26157 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9UG22 | |
GIMAP2 | |
A synthesized peptide derived from human GIMAP2(Accession Q9UG22), corresponding to amino acid residues E208-E258. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction