Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GIMAP3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988418
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8418 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8418 Supplier Bioss Supplier No. BS8269R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GIMAP3p is a murine mitochondrial membrane protein belonging to the GTP-binding superfamily and to the immuno-associated nucleotide (IAN) subfamily of nucleotide-binding proteins. The IAN/GIMAP family consists of GTP-binding proteins that share a unique primary structure and whose expression is finely regulated by T-cell receptor signals. Recent studies have shown that IAN/GIMAP family proteins crucially regulate the survival of T cells during development, selection and homeostasis, and are possibly linked to the onset of T-lymphopenia, leukemia and autoimmunity. IAN/GIMAP family proteins might also take part in mitochondrial regulation of lymphocyte apoptosis by interacting with Bcl-2 family proteins. The human GIMAP (GTPase of the immunity-associated protein) gene family includes seven functional members residing on (human) chromosome 7q36.1 and one pseudogene (hGIMAP3).

Specifications

Antigen GIMAP3
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene LOC105370575
Gene Symbols LOC105370575
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from mouse GIMAP3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 105370575
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.