Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GIMAP4 Antibody (OTI1C6), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271997PE
Description
GIMAP4 Monoclonal specifically detects GIMAP4 in Human samples. It is validated for Flow Cytometry.Specifications
GIMAP4 | |
Monoclonal | |
PE | |
PBS with 0.05% Sodium Azide | |
GIMAP4 | |
Full length recombinant protein of human GIMAP4 (NP_060796) produced in HEK293T cells. | |
0.1 mL | |
55303.0 | |
Store at 4°C in the dark. |
Flow Cytometry | |
OTI1C6 | |
Flow Cytometry | |
GTPase IMAP family member 4, GTPase, IMAP family member 4, HIMAP4, human immune associated nucleotide 1, IAN-1, IAN1FLJ11110, IMAP4hIAN1, immunity associated protein 4, Immunity-associated nucleotide 1 protein, Immunity-associated protein 4 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction