Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GIMAP4 Monoclonal Antibody (OTI1A11)
GREENER_CHOICE

Catalog No. PIMA525982
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525982 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525982 Supplier Invitrogen™ Supplier No. MA525982
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GIMAP4 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

This gene encodes a protein belonging to the GTP-binding superfamily and to the immuno-associated nucleotide (IAN) subfamily of nucleotide-binding proteins. The encoded protein of this gene may be negatively regulated by T-cell acute lymphocytic leukemia 1 (TAL1). In humans, the IAN subfamily genes are located in a cluster at 7q36.1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GIMAP4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI1A11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GIMAP4
Gene Accession No. Q9NUV9
Gene Alias AU019574; E430007K16Rik; GIMAP4; GTPase IMAP family member 4; GTPase, IMAP family member 4; human immune associated nucleotide 1; Ian1; IAN-1; Imap4; immune associated nucleotide 1; immune-associated nucleotide 1; immune-associated nucleotide-binding protein 1; immunity associated protein 4; immunity-associated nucleotide 1 protein; Immunity-associated protein 4; MSTP062
Gene Symbols GIMAP4
Host Species Mouse
Immunogen Full length human recombinant protein of GIMAP4 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55303
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.