Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GIMAP4 Mouse anti-Human, Biotin, Clone: OTI2F9, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA700481

Description
This gene encodes a protein belonging to the GTP-binding superfamily and to the immuno-associated nucleotide (IAN) subfamily of nucleotide-binding proteins. The encoded protein of this gene may be negatively regulated by T-cell acute lymphocytic leukemia 1 (TAL1). In humans, the IAN subfamily genes are located in a cluster at 7q36.1.Specifications
GIMAP4 | |
OTI2F9 | |
Biotin | |
GIMAP4 | |
IAN-1, IAN1, IMAP4, MSTP062 | |
Mouse | |
Affinity Chromatography | |
RUO | |
55303 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Monoclonal | |
0.5 mg/mL | |
PBS with 10mg/ml BSA, 50% glycerol and 0.05% sodium azide | |
Q9NUV9 | |
GIMAP4 | |
Full length human recombit protein of human GIMAP4 produced in HEK293T cell. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction