Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GIMAP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555540
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555540 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555540 Supplier Thermo Scientific Supplier No. PA555540
Add to Cart
Add to Cart

GIMAP4 Polyclonal Antibody

GIMAP4 Polyclonal Antibody for Western Blot, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen GIMAP4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GIMAP4
Gene Accession No. Q9NUV9
Gene Alias AU019574; E430007K16Rik; GIMAP4; GTPase IMAP family member 4; GTPase, IMAP family member 4; human immune associated nucleotide 1; Ian1; IAN-1; Imap4; immune associated nucleotide 1; immune-associated nucleotide 1; immune-associated nucleotide-binding protein 1; immunity associated protein 4; immunity-associated nucleotide 1 protein; Immunity-associated protein 4; MSTP062
Gene Symbols GIMAP4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GIMAP4. Recombinant protein control fragment (Product #RP-92507).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55303
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.