Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GIMAP5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA528872
Description
Recommended positive controls: HeLaS3. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a protein belonging to the GTP-binding superfamily and to the immuno-associated nucleotide (IAN) subfamily of nucleotide-binding proteins. In humans, the IAN subfamily genes are located in a cluster at 7q36.1.
Specifications
| GIMAP5 | |
| Polyclonal | |
| Unconjugated | |
| GIMAP5 | |
| D630024P16; E230026N22Rik; Gimap5; GTPase IMAP family member 5; GTPase of the immunity-associated protein 5; GTPase, IMAP family member 5; HIMAP3; Ian4; IAN-4; Ian4l1; IAN5; IAN-5; Iddm1; IMAP3; immune associated nucleotide 4 like 1; immune-associated nucleotide-binding protein 5; immunity associated protein 3; immunity-associated nucleotide 4 protein; Immunity-associated nucleotide 4-like 1 protein; immunity-associated nucleotide 5 protein; Immunity-associated protein 3; inhibitor of radiation- and OA-induced apoptosis, Irod/Ian5; IROD; lymphopenia; lyp | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55340 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q96F15 | |
| GIMAP5 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human GIMAP5. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction