Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GIMAP5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550042
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550042 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550042 Supplier Invitrogen™ Supplier No. PA550042
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total GIMAP5 protein.

This gene encodes a protein belonging to the GTP-binding superfamily and to the immuno-associated nucleotide (IAN) subfamily of nucleotide-binding proteins. In humans, the IAN subfamily genes are located in a cluster at 7q36.1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GIMAP5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GIMAP5
Gene Accession No. Q96F15
Gene Alias D630024P16; E230026N22Rik; Gimap5; GTPase IMAP family member 5; GTPase of the immunity-associated protein 5; GTPase, IMAP family member 5; HIMAP3; Ian4; IAN-4; Ian4l1; IAN5; IAN-5; Iddm1; IMAP3; immune associated nucleotide 4 like 1; immune-associated nucleotide-binding protein 5; immunity associated protein 3; immunity-associated nucleotide 4 protein; Immunity-associated nucleotide 4-like 1 protein; immunity-associated nucleotide 5 protein; Immunity-associated protein 3; inhibitor of radiation- and OA-induced apoptosis, Irod/Ian5; IROD; lymphopenia; lyp
Gene Symbols GIMAP5
Host Species Rabbit
Immunogen Synthesized peptide derived from internal of human GIMAP5.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55340
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.