Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GIMAP6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584028
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584028 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584028 Supplier Invitrogen™ Supplier No. PA584028
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SLEDYVRETN NQALAWLDVT LARRHCGFNN RAQGEEQEAQ LRELMEKVEA IMWENEGDYY SNKAYQYTQQ NFRLKELQER QVSQGQGSED VPGEESWLEG LSQIQK Highest antigen sequence indentity to the following orthologs: Rat - 38%, Mouse - 38%.

This gene encodes a protein belonging to the GTP-binding superfamily and to the immuno-associated nucleotide (IAN) subfamily of nucleotide-binding proteins. In humans, IAN subfamily genes are located in a cluster at 7q36.1. Two transcript variants, one protein-coding and the other probably non-protein-coding, have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GIMAP6
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GIMAP6
Gene Accession No. Q6P9H5
Gene Alias 4833419H03Rik; Gimap6; GTPase IMAP family member 6; GTPase, IMAP family member 6; IAN2; IAN-2; IAN6; IAN-6; immune associated nucleotide 2; immune associated nucleotide 6; immune-associated nucleotide 6; immune-associated nucleotide-binding protein 6; Immunity-associated nucleotide 2 protein; Immunity-associated nucleotide 6 protein; mFLJ00102; mIAN6
Gene Symbols GIMAP6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GIMAP6. Recombinant protein control fragment (Product #RP-106307).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 474344
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.