Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GIMAP6 Rabbit anti-Human, Alexa Fluor 350, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306038AF350
Description
GIMAP6 Polyclonal specifically detects GIMAP6 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
GIMAP6 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
DKFZp686A01175, FLJ22690, GTPase IMAP family member 6, GTPase, IMAP family member 6, hIAN2, hIAN6, human immune associated nucleotide 2, IAN2, IAN-2, IAN6, IAN-6, immune associated nucleotide 2, immune associated nucleotide 6, Immunity-associated nucleotide 2 protein, Immunity-associated nucleotide 6 protein | |
Produced in rabbits immunized with E. coli-derived Human GIMAP6 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
474344 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction