Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GIMAP7 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | GIMAP7 |
---|---|
Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:200-1:500 |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
GIMAP7 Polyclonal specifically detects GIMAP7 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
GIMAP7 | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
168537 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:MAESEDRSLRIVLVGKTGSGKSATANTILGEEIFDSRIAAQAVTKNCQKASREWQGRDLLVVDT | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot 0.4 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:200-1:500 | |
Polyclonal | |
Rabbit | |
Human | |
GTPase, IMAP family member 7, hIAN7, IAN-7, IAN7GTPase IMAP family member 7, immune associated nucleotide, Immunity-associated nucleotide 7 protein, MGC27027 | |
GIMAP7 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title