Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GIMAP7 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988422
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8422 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8422 Supplier Bioss Supplier No. BS8273R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GIMAP7 is also name as hIAN7 or IAN 7. The function of GIMAP7 remains largely unknown.

Specifications

Antigen GIMAP7
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GIMAP7
Gene Accession No. Q8NHV1
Gene Alias GIMAP7; GTPase IMAP family member 7; GTPase, IMAP family member 7; hIAN7; IAN7; IAN-7; immune associated nucleotide; immune-associated nucleotide-binding protein 7; Immunity-associated nucleotide 7 protein
Gene Symbols GIMAP7
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GIMAP7.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 168537
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.