Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GIMAP8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553641
Description
Immunogen sequence: SWVLRERKVV VIDTPDLFSS IACAEDKQRN IQHCLELSAP SLHALLLVIA IGHFTREDEE TAKGIQQVFG AEARRHIIIV FTRKDDLGDD LLQDFIEKNK PLKQLVQDYE GRYCIFNNKT NSKDEQITQV LELLRKVESL VNT Highest antigen sequence identity to the following orthologs: Mouse - 46%, Rat - 48%.

Specifications
GIMAP8 | |
Polyclonal | |
Unconjugated | |
GIMAP8 | |
GIMAP8; Gm457; GTPase IMAP family member 8; GTPase, IMAP family member 8; hIAN6; human immune associated nucleotide 6; IAN6; IAN9; IAN-9; Iant; IMAP8; Immune-associated nucleotide-binding protein 9; immunity-associated protein 8; mGIMAP8; Protein IanT | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
155038 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8ND71 | |
GIMAP8 | |
Recombinant protein corresponding to Human GIMAP8. Recombinant protein control fragment (Product #RP-90834). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction