Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GIMAP8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590299
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590299 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590299 Supplier Invitrogen™ Supplier No. PA590299
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSEQSCQMSE LRLLLLGKCR SGKSATGNAI LGKHVFKSKF SDQTVIKMCQ RESWVLRERK VVVIDTPDLF SSIACAEDKQ RNIQHCLELS APSLHALLLV IAIGHFTRED EETAKGIQQV FGAEARRHII IVFTRKDDLG DDLLQDFIEK NKPLKQLVQD YEGRYCIFNN KTNSKDEQIT QVLELLRKVE SLVNTNGGPY HVNFKTEGSR FQDCVNEAAS QEGDKPQGPR ERQLQSTGPE QNPGTSELTV LLVGKRGAGK SAAGNSILGR QAFQTGFSEQ SVTQSFLSES RSWRKKKVSI; Positive Samples: A549, mouse thymus, mouse spleen, rat spleen; Cellular Location: Cytoplasm, Endoplasmic reticulum, Golgi apparatus, Mitochondrion, cytosol.

The GIMAP8 gene, part of the GTPase of the Immunity-Associated Proteins (GIMAP) family, is located on chromosome 7q36.1. This gene encodes a protein that is predominantly expressed in immune cells, including T and B lymphocytes. GIMAP8 is involved in the regulation of lymphocyte survival and development, particularly during the early and late stages of T cell development in the thymus, as well as during B cell maturation. Studies have shown that GIMAP8 expression levels are significantly altered in various cancers, including breast and lung adenocarcinoma, where its upregulation in tumor tissues correlates with improved overall survival in specific cancer subtypes. This suggests a potential role for GIMAP8 in tumor immune surveillance and as a prognostic biomarker. Additionally, GIMAP8's functions appear essential but redundant enough that mouse models deficient in this gene do not exhibit major defects in T or B cell development, indicating compensation by other family members.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GIMAP8
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene GIMAP8
Gene Accession No. Q4KLG2, Q75N62, Q8ND71
Gene Alias GIMAP8; Gm457; GTPase IMAP family member 8; GTPase, IMAP family member 8; hIAN6; human immune associated nucleotide 6; IAN6; IAN9; IAN-9; Iant; IMAP8; Immune-associated nucleotide-binding protein 9; immunity-associated protein 8; mGIMAP8; Protein IanT
Gene Symbols GIMAP8
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human GIMAP8 (NP_7831611).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 155038, 243374, 500112
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.