Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GIMAP8 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA521969

Catalog No. PIPA521969


Only null left
Add to Cart

Description

Description

Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The GIMAP8 gene, part of the GTPase of the Immunity-Associated Proteins (GIMAP) family, is located on chromosome 7q36.1. This gene encodes a protein that is predominantly expressed in immune cells, including T and B lymphocytes. GIMAP8 is involved in the regulation of lymphocyte survival and development, particularly during the early and late stages of T cell development in the thymus, as well as during B cell maturation. Studies have shown that GIMAP8 expression levels are significantly altered in various cancers, including breast and lung adenocarcinoma, where its upregulation in tumor tissues correlates with improved overall survival in specific cancer subtypes. This suggests a potential role for GIMAP8 in tumor immune surveillance and as a prognostic biomarker. Additionally, GIMAP8's functions appear essential but redundant enough that mouse models deficient in this gene do not exhibit major defects in T or B cell development, indicating compensation by other family members.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GIMAP8
Polyclonal
Unconjugated
GIMAP8
GIMAP8; Gm457; GTPase IMAP family member 8; GTPase, IMAP family member 8; hIAN6; human immune associated nucleotide 6; IAN6; IAN9; IAN-9; Iant; IMAP8; Immune-associated nucleotide-binding protein 9; immunity-associated protein 8; mGIMAP8; Protein IanT
Rabbit
Antigen affinity chromatography
RUO
155038
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q8ND71
GIMAP8
Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human GIMAP8. The exact sequence is proprietary.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.