Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GIMAP8 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA521970

Catalog No. PIPA521970


Only null left
Add to Cart

Description

Description

Recommended positive controls: 293T, A431, Jurkat, Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

GIMAP8
Polyclonal
Unconjugated
GIMAP8
GIMAP8; Gm457; GTPase IMAP family member 8; GTPase, IMAP family member 8; hIAN6; human immune associated nucleotide 6; IAN6; IAN9; IAN-9; Iant; IMAP8; Immune-associated nucleotide-binding protein 9; immunity-associated protein 8; mGIMAP8; Protein IanT
Rabbit
Antigen Affinity Chromatography
RUO
155038
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q8ND71
GIMAP8
Synthetic peptide corresponding to a region within amino acids 352 and 443 of GIMAP8 (Uniprot ID#Q8ND71).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.