Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GIN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566381
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566381 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566381 Supplier Invitrogen™ Supplier No. PA566381
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WKDGRFQSEWV GPCVIDYITE SGCAVLRDNT GVRLKRPIKM SHLKPYIRES SEQESLYLLQ GSVVADHDYI GLPEIPIGA Highest antigen sequence identity to the following orthologs - mouse 91%, rat 89%.

GIN1 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include nucleic acid binding; DNA integration.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GIN1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GIN1
Gene Accession No. Q9NXP7
Gene Alias 4930429M06Rik; 8430411H10Rik; Gin1; Gin-1; gypsy retrotransposon integrase 1; gypsy retrotransposon integrase-like protein 1; TGIN1; Ty3/Gypsy integrase 1; ZH2C2; Zinc finger H2C2 domain-containing protein; zinc finger, H2C2 domain containing
Gene Symbols GIN1
Host Species Rabbit
Immunogen Recombinant Human GIN1. Recombinant protein control fragment (Product #RP-107056).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54826
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.