Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GIN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571571
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571571 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571571 Supplier Invitrogen™ Supplier No. PA571571
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GIN1 Polyclonal Antibody for IHC (P), Western Blot

GIN1 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include nucleic acid binding; DNA integration.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GIN1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene GIN1
Gene Accession No. Q8K259, Q9NXP7
Gene Alias 4930429M06Rik; 8430411H10Rik; Gin1; Gin-1; gypsy retrotransposon integrase 1; gypsy retrotransposon integrase-like protein 1; TGIN1; Ty3/Gypsy integrase 1; ZH2C2; Zinc finger H2C2 domain-containing protein; zinc finger, H2C2 domain containing
Gene Symbols GIN1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 464-492 amino acids from the C-terminal region of human GIN1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 252876, 54826
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.