Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GINM1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559671
Description
Immunogen sequence: QKDVTEIDIL VKNRGVLRHS NYTLPLEESM LYSISRDSDI LFTLPNLSKK ESVSSLQTTS QYLIRNVETT VDEDVLPGKL PETPLRAEPP SSYKVMCQWM EKFRKDLCRF WSN Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 75%.

Specifications
GINM1 | |
Polyclonal | |
Unconjugated | |
GINM1 | |
AA589616; C6orf72; dJ12G14.2; Ginm1; glycoprotein integral membrane 1; glycoprotein integral membrane protein 1; RGD1309759; UNQ710/PRO1361 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
116254 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NU53 | |
GINM1 | |
Recombinant protein corresponding to Human GINM1. Recombinant protein control fragment (Product #RP-102913). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction