Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GINS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562341
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562341 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562341 Supplier Invitrogen™ Supplier No. PA562341
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HMAAEEMEWF NNYKRSLATY MRSLGGDEGL DITQDMKPPK SLYIEVRCLK DYGEFEVDDG TSVLLKKNSQ HFLPRWKCEQ LIRQGVLEH Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.

The yeast heterotetrameric GINS complex is made up of Sld5 (GINS4; MIM 610611), Psf1, Psf2 (GINS2; MIM 610609), and Psf3 (GINS3; MIM 610610). The formation of the GINS complex is essential for the initiation of DNA replication in yeast and Xenopus egg extracts.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GINS1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GINS1
Gene Accession No. Q14691
Gene Alias 2810418N01Rik; DNA replication complex GINS protein PSF1; GINS complex subunit 1; GINS complex subunit 1 (Psf1 homolog); GINS complex subunit 4 (Sld5 homolog); Gins1; Gins4; KIAA0186; mKIAA0186; partner of sld five-1; PSF1; RP4-691N24.2
Gene Symbols GINS1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GINS1. Recombinant protein control fragment (Product #RP-99313).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9837
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.