Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GINS2 Monoclonal Antibody (3H4)
GREENER_CHOICE

Catalog No. PIMA521970
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA521970 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA521970 Supplier Thermo Scientific Supplier No. MA521970
Add to Cart
Add to Cart

GINS2 Monoclonal Antibody (3H4)

GINS2 Monoclonal Antibody for Western Blot, ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen GINS2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3H4
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene GINS2
Gene Accession No. Q9Y248
Gene Alias 2210013I18Rik; 4833427B12Rik; AI323585; CGI-122; DC5; DNA replication complex GINS protein PSF2; GINS complex subunit 2; GINS complex subunit 2 (Psf2 homolog); Gins2; HSPC037; Pfs2; PSF2; RGD1311055
Gene Symbols GINS2
Host Species Mouse
Immunogen GINS2 (AAH03186, 1 a.a. ~ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51659
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.