Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GINS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542838
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542838 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542838 Supplier Invitrogen™ Supplier No. PA542838
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: VRQQEAHAKL DNLTLMEINT SGTFLTQALN HMYKLRTNLQ PLESTQSQDF Sequence homology: Cow: 79%; Dog: 93%; Horse: 86%; Human: 100%; Mouse: 93%; Rat: 86%.

The yeast heterotetrameric GINS complex is made up of Sld5, Psf1, Psf2, and Psf3. The formation of this complex is essential for the initiation of DNA replication in yeast and Xenopus egg extracts (Ueno et al., 2005 ). See GINS1 for additional information about the GINS complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GINS2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GINS2
Gene Accession No. Q9Y248
Gene Alias 2210013I18Rik; 4833427B12Rik; AI323585; CGI-122; DC5; DNA replication complex GINS protein PSF2; GINS complex subunit 2; GINS complex subunit 2 (Psf2 homolog); Gins2; HSPC037; Pfs2; PSF2; RGD1311055
Gene Symbols GINS2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human GINS2 (aa 136-185).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51659
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.